missing translation for 'onlineSavingsMsg'
Learn More

Potassium Channel Kv2.1 Rabbit anti-Rat, Polyclonal, MilliporeSigma™

Catalog No. AB518650UL
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB518650UL 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB518650UL Supplier MilliporeSigma Supplier No. AB518650UL
Only null left

Rabbit Polyclonal Antibody

Anti-Potassium Channel Kv2.1 Antibody detects level of Potassium Channel Kv2.1 and has been published and validated for use in immunohistochemistry and western blotting.

Specifications

Antigen Potassium Channel Kv2.1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified immunoglobulin by Protein A. Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA, 5% sucrose as a stabilizer and 0.025% sodium azide as a preservative. Reconstitute with 50μL of sterile deionized water. Centrifuge antibody
Gene Symbols KCNB1; KV2.1; h-DRK1; Kv2.1; DRK1
Host Species Rabbit
Immunogen Purified peptide from rat Kv2.1 (amino acids 841-857) (Accession P15387)
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_004975.2
Target Species Rat
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only