missing translation for 'onlineSavingsMsg'
Learn More

Podocalyxin-like protein I (Cytotoxic) Mouse anti-Human, Clone: mab 84, MilliporeSigma™

Catalog No. MABD89MI
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABD89MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABD89MI Supplier MilliporeSigma Supplier No. MABD89
Only null left

Mouse Monoclonal Antibody

Anti-Podocalyxin-like protein I (Cytotoxic) Antibody, clone mAb 84 is an antibody against Podocalyxin-like protein I (Cytotoxic) for use in Western Blotting, immunocytochemistry, Flow Cytometry.

Specifications

Antigen Podocalyxin-like protein I (Cytotoxic)
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone mab 84
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgMκ in buffer containing PBS without preservatives.
Gene Accession No. NP_001018121
Gene Symbols PCLP, Gp200, PC, PCLP1
Host Species Mouse
Immunogen HES-3 Human embryonic stem cell line
Purification Method Purified by ion-exchange chromatography
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgM κ
Show More Show Less

For Research Use Only