missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PNMA2, Mouse, Clone: 4F10, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant PNMA2.
Supplier: Abnova Corporation H00010687M10
Description
Sequence: MALALLEDWCRIMSVDEQKSLMVTGIPADFEEAEIQEVLQETLKSLGRYRLLGKIFRKQENANAVLLELLEDTDVSAIPSEVQGKGGVWKVIFKTPNQDTEFLERLNLFLEKEGQTVSGMFRALGQEGVSPATVPCISPELLAHLLGQAMAHAPQPLLPMRYRKLRVFSGSAVPAPEEESFEVWLEQATEIVKEWPVTEAEKKRWLAESLRGPALDLMHIVQADNPSISVEECLEAFKQVFGSLESRRTAQVRYLKTYQEEGEKVSAYVLRLETLLRRAVEKRAIPRRIADQVRLEQVMAGATLNQMLWCRLRELKDQGPPPSFLELMKVIREEEEEEASFENESIEEPEERDGYGRWNHEGDDSpecifications
| PNMA2, | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC062301 | |
| PNMA2 | |
| PNMA2 (AAH62301, 1 a.a. ∼ 364 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 4F10 | |
| Mouse monoclonal antibody raised against a full-length recombinant PNMA2. | |
| PNMA2 | |
| KIAA0883/MA2/MM2/RGAG2 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10687 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |