missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plexin-A3 Mouse anti-Rat, Human, Mouse, Clone: 18A9.2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB10422
Description
Anti-Plexin-A3 Antibody, clone 18A9.2 is a Mouse Monoclonal Antibody for detection of Plexin-A3 also known as PLEXIN-A3, Semaphorin receptor SEX or Plexin-4 and has been validated in western blotting and immunohistochemistry.Specifications
| Plexin-A3 | |
| Monoclonal | |
| Unconjugated | |
| NP_059984 | |
| Mouse | |
| Protein G | |
| Primary | |
| 2-8°C for one year | |
| IgG1 κ |
| Immunohistochemistry, Western Blot | |
| 18A9.2 | |
| Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
| HSSEXGENE, SEX, XAP-6, PLXN3, PLXN4, Plxn3, PLXNA3 | |
| GST-tagged recombinant protein corresponding to human Plexin-A3. | |
| 100 μg | |
| Human, Mouse, Rat | |
| Purified |
For Research Use Only