missing translation for 'onlineSavingsMsg'
Learn More

PLEKHG4B Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. 072143MI
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
072143MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 072143MI Supplier MilliporeSigma Supplier No. JUL43
Only null left

Rabbit Polyclonal Antibody

Detect Pleckstrin homology domain-containing family G member 4B using this rabbit polyclonal antibody, Anti-PLEKHG4B Antibody validated for use in western blotting.

Specifications

Antigen PLEKHG4B
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols PLEKHG4B; KIAA1909
Host Species Rabbit
Immunogen GST-tagged recombinant protein corresponding to human PLEKHG4B.
Purification Method Affinity Purified
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_443141
Target Species Human
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only