Learn More
phospholipase D2, Mouse, Clone: 1C5, Abnova™
Mouse monoclonal antibody raised against a partial recombinant PLD2.
Supplier: Abnova Corporation H00005338M01
Description
Phosphatidylcholine (PC)-specific phospholipases D (PLDs; EC 3.1.4.4) catalyze the hydrolysis of PC to produce phosphatidic acid and choline. Activation of PC-specific PLDs occurs as a consequence of agonist stimulation of both tyrosine kinase and G protein-coupled receptors. PC-specific PLDs have been proposed to function in regulated secretion, cytoskeletal reorganization, transcriptional regulation, and cell cycle control.[supplied by OMIM
Sequence: LRDPICDDFFQLWQDMAESNANIYEQIFRCLPSNATRSLRTLREYVAVEPLATVSPPLARSELTQVQGHLVHFPLKFLEDESLLPPLGSKEGMIPLEVWTSpecifications
| phospholipase D2 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC015033 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Antibody |
| ELISA, Western Blot | |
| 1C5 | |
| Mouse monoclonal antibody raised against a partial recombinant PLD2. | |
| PLD2 | |
| PLD2 | |
| PLD2 (AAH15033, 834 a.a. ∼ 933 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Signal Transduction | |
| 5338 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |