Learn More
phospholipase A2, group IB (pancreas), Mouse, Clone: 2B7, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant PLA2G1B.
Supplier: Abnova Corporation H00005319M14
Description
Phospholipase A2 (EC 3.1.1.4) catalyzes the release of fatty acids from glycero-3-phosphocholines. The best known varieties are the digestive enzymes secreted as zymogens by the pancreas of mammals. Sequences of pancreatic PLA2 enzymes from a variety of mammals have been reported. One striking feature of these enzymes is their close homology to venom phospholipases of snakes. Other forms of PLA2 have been isolated from brain, liver, lung, spleen, intestine, macrophages, leukocytes, erythrocytes, inflammatory exudates, chondrocytes, and platelets (Seilhamer et al., 1986 [PubMed 3028739]) .[supplied by OMIM
Sequence: SGISPRAVWQFRKMIKCVIPGSDPFLEYNNYGCYCGLGGSGTPVDELDKQKQRVSpecifications
| phospholipase A2, group IB (pancreas) | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC005386 | |
| PLA2G1B | |
| PLA2G1B (AAH05386, 17 a.a. ∼ 70 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Endocrine/Metabolism | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 2B7 | |
| Mouse monoclonal antibody raised against a full length recombinant PLA2G1B. | |
| PLA2G1B | |
| MGC119834/MGC119835/PLA2/PLA2A/PPLA2 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 5319 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |