missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phosphoinositide-3-kinase, regulatory subunit 4, Mouse, Clone: 1B5, Abnova™
Mouse monoclonal antibody raised against a partial recombinant PIK3R4.
Supplier: Abnova Corporation H00030849M02
Description
Sequence: AGSDMKIRFWDLAYPERSYVVAGSTSSPSVSYYRKIIEGTEVVQEIQNKQKVGPSDDTPRRGPESLPVGHHDIITDVATFQTTQGFIVTASRDGIVKVWKSpecifications
| phosphoinositide-3-kinase, regulatory subunit 4 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_014602 | |
| PIK3R4 | |
| PIK3R4 (NP_055417, 1259 a.a. ∼ 1358 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Kinases | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| 1B5 | |
| Mouse monoclonal antibody raised against a partial recombinant PIK3R4. | |
| PIK3R4 | |
| MGC102700/VPS15/p150 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 30849 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |