missing translation for 'onlineSavingsMsg'
Learn More

Phospho-tyrosine Mouse anti-Vertebrates, Clone: 4G10™, MilliporeSigma™

Catalog No. 05321XMI
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05321XMI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05321XMI Supplier MilliporeSigma Supplier No. 05321X
Only null left

Mouse Monoclonal Antibody

Anti-Phosphotyrosine Antibody, clone 4G10 detects tyrosine phosphorylated proteins in all species. This unique monoclonal antibody is validated for use in immunocytochemistry, immunohistochemistry, immunoprecipitation, western blotting and is backed by hundreds of publications.

Specifications

Antigen Phosphotyrosine
Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 4G10™
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2bκ in buffer containing 0.1MTris-glycine, pH 7.4, 0.15MNaCl, 0.05% sodium azide. Liquid at 2-8ºC.
IgG2bκ mouse monoclonal antibody produced in vitro by mouse-mouse hybridoma 4G10™ (FOX-NY [NS-1 derivative]
Host Species Mouse
Immunogen Phosphotyramine-KLH
Purification Method Protein G-Sepharose™ chromatography
Quantity 50 μg
Primary or Secondary Primary
Target Species Vertebrates
Content And Storage 2-8°C for 12 months. Do not freeze.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only