missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-tyrosine Mouse anti-All, Clone: 4G10™, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05321
Description
Anti-Phosphotyrosine Antibody, clone 4G10 detects tyrosine phosphorylated proteins in all species. This unique monoclonal antibody is validated for use in immunocytochemistry, immunohistochemistry, immunoprecipitation, western blotting and is backed by hundreds of publicationsSpecifications
| Phosphotyrosine | |
| Monoclonal | |
| Unconjugated | |
| Mouse | |
| Protein G-Sepharose™ chromatography | |
| Primary | |
| 2-8°C for 12 months. Do not freeze. | |
| IgG2b κ |
| Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 4G10™ | |
| 0.1M Tris-Glycine, 0.15M NaCl, 0.05% Sodium Azide, pH 7.4. Liquid at 2-8°C. IgG2bκ mouse monocolonal antibody produced in vitro by mouse-mouse hybridoma 4G10™ (FOX-NY [NS-1 derivative] myeloma x spleen cells). | |
| Phosphotyramine-KLH | |
| 100 μg | |
| All species | |
| Purified |
For Research Use Only