missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-serine Rabbit anti-All, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB1603
Description
Anti-Phosphoserine Antibody detects level of Phosphoserine and has been published and validated for use in ELISA, immunohistochemistry, immunoprecipitation and western blotting.Specifications
| Phosphoserine | |
| Polyclonal | |
| Purified rabbit in buffer containing 50% glycerol. | |
| Keyhole Limpet Hemocyanin(KLH)- conjugated phosphoserine conjugates. | |
| 50 μg | |
| All species |
| ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Affinity Purified | |
| Primary | |
| -20°C in undiluted aliquotes for up to 6 months, Avoid Freeze/Thaw Cycles |
For Research Use Only