missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-STAT5A/B (Ser726/Ser731) Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 06867
Description
Anti-phospho-STAT5A/B (Ser726/Ser731) Antibody is a rabbit polyclonal antibody for detection of phospho-STAT5A/B (Ser726/Ser731) also known as signal transducer and activator of transcription 5A/B and has been tested in western blotting.Specifications
| phospho-STAT5A/B (Ser726/Ser731) | |
| Polyclonal | |
| rabbit antiserum in 30% glycerol and 0.05% sodium azide | |
| Rabbit | |
| Antiserum | |
| Primary | |
| Human, Mouse, Rat | |
| Serum |
| Western Blot | |
| Unconjugated | |
| STAT5A; MGF; STAT5 | |
| peptide (DQAP[pS]PAVC) corresponding to amino acids 722-730 of human STAT5A or 727-735 of human STAT5B | |
| 200 μL | |
| NM_003152.2 | |
| −20°C for 24 months | |
| IgG |
For Research Use Only