missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-PLK1 (Ser137) Rabbit anti-Human, Mouse, Rat, Bovine, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 071348
Description
Detect phospho-PLK1 (Ser137) using this Anti-phospho-PLK1 (Ser137) Antibody validated for use in western blotting.Specifications
| phospho-PLK1 (Ser137) | |
| Polyclonal | |
| Affinity purified rabbit serum in 0.1M Tris-Glycine (pH7.4), 150mM NaCl with 0.05% sodium azide. | |
| Rabbit | |
| Affinity Purified | |
| Primary | |
| Bovine, Human, Mouse, Rat |
| Western Blot | |
| Unconjugated | |
| EC 2.7.11.21; PLK ; PLK-1; STPK13 | |
| KLH-conjugated linear peptide corresponding to human phosphoylated PLK1 at and around Ser137. | |
| 100 μg | |
| NP_005021 | |
| 2-8°C for 12 months |
For Research Use Only