missing translation for 'onlineSavingsMsg'
Learn More

Phospho-PKR (Thr451) Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 07886MI
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07886MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07886MI Supplier MilliporeSigma Supplier No. MIL07886
Only null left

Rabbit Polyclonal Antibody

This Anti-phospho-PKR (Thr451) Antibody is validated for use in western blotting for the detection of phospho-PKR (Thr451).

Specifications

Antigen phospho-PKR (Thr451)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 100μL of affinity purified rabbit polyclonal IgG in PBS (without Mg2+ and Ca2+), pH 7.3 containing 0.5mg/mL BSA (IgG, protease free) and 0.025% sodium azide and 50% glycerol.
Gene Symbols EIF2AK2; PRKR; PKR; MGC126524; EIF2AK1
Host Species Rabbit
Immunogen Peptide corresponding to amino acid region encompassing the human phospho-PKR (Thr451)
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_002759.1
Target Species Human, Mouse
Content And Storage -20°C in undiluted aliquotes for up to 24 months, Avoid Freeze/Thaw Cycles
Isotype IgG
Show More Show Less

For Research Use Only