missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Phospho-MKK7/SKK4 (Thr275/Ser277) Rabbit anti-Human, Polyclonal
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma™ 36010
Description
Anti-Phospho-MKK7/SKK4 (Thr275/Ser277), Cat. No. 36-010, is a highly specific rabbit polyclonal antibody that detect dual specificity mitogen-activated protein kinase kinase 7 (MKK7) and has been tested for use in Western Blotting.Specifications
| phospho-MKK7/SKK4 (Thr275/Ser277) | |
| Polyclonal | |
| 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%. | |
| Rabbit | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG |
| Western Blot | |
| Unconjugated | |
| MAP2K7; MKK7; PRKMK7; MEK7; MAPKK7; JNKK2; Jnkk2 | |
| peptide corresponding to amino acids surrounding Thr275/Ser277 of human MKK7/SKK4. | |
| 100 μL | |
| NM_145185.2 | |
| −20°C for 12 months. |
For Research Use Only