missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-JAK-2 (Tyr570) Rabbit anti-Human, Rat, Horse, Chimpanzee, Pig, Primate, Polyclonal, MilliporeSigma™
Description
This Anti-phospho JAK-2 (Tyr570) Antibody is validated for use in western blot, western blot, western blotting for the detection of phospho JAK-2 (Tyr570).
Specifications
Specifications
| Antigen | phospho JAK-2 (Tyr570) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide. |
| Gene Symbols | JAK-2; JTK10 |
| Host Species | Rabbit |
| Immunogen | KLH-conjugated linear peptide corresponding to human JAK-2 phosphorylated at Tyr570. |
| Purification Method | Affinity Purified |
| Quantity | 100 μg |
| Show More |
For Research Use Only