missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Histone H3 (Thr11) Mouse anti-Human, Clone: N123/48, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABE435
Description
This Anti-phospho Histone H3 (Thr11) Antibody, clone N123/48 is validated for use in western blotting, immunocytochemistry and dot blot for the detection of phospho Histone H3 (Thr11).Specifications
| phospho Histone H3 (Thr11) | |
| Monoclonal | |
| 0.5mg/mL | |
| Purified mouse monoclonal IgG1 in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
| Mouse | |
| Protein G purified | |
| Primary | |
| 2-8°C for one year | |
| IgG1 |
| Dot Blot, Immunocytochemistry, Western Blot | |
| N123/48 | |
| Unconjugated | |
| NP_001013074 | |
| Linear peptide corresponding to rat Histone H3 phosphorylated at Thr11. | |
| 100 μL | |
| Human | |
| Purified |
For Research Use Only