missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Phospho-Histone H2A.X (Ser139) Mouse anti-Human, FITC, Clone: JBW301
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ 16202A
Description
Use Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, FITC conjugate (mouse monoclonal antibody) validated in flow cytometry, immunocytochemistry to detect phospho-Histone H2A.X (Ser139) also known as H2AXS139P, H2AX histone.Specifications
| phospho-Histone H2A.X (Ser139) | |
| Monoclonal | |
| FITC | |
| NM_002105.2 | |
| Mouse | |
| Protein G purfied | |
| Primary | |
| Stable for 1 year at from date of receipt. | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry | |
| JBW301 | |
| Purified FITC-conjugated mouse monoclonal IgG1 in buffer containing PBS with 0.05% sodium azide. Frozen solution. | |
| H2AFX, H2AX, H2a/x, H2A/X, H2A.X | |
| KLH-conjugated, synthetic peptide (CKATQA[pS]QEY) corresponding to amino acids 134-142 of human histone H2A.X. The immunizing sequence has 8 identical amino acids in yeast and mouse. Clone JBW301. | |
| 100 μg | |
| Human | |
| Purified |
For Research Use Only