missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-H2A.X (Ser139) Rabbit anti-Human, AlexaFluor™647, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 07164AF647
Description
This AlexaFluor™647 conjugated Anti-phospho Histone H2A.X antibody is validated for use in mmunocytochemistry for the detection of phospho-Histone H2A.X.Specifications
| phospho H2A.X (Ser139) | |
| Polyclonal | |
| Purified conjugated rabbit polyclonal antibody in buffer containing PBS and 15mg/mL BSA with 0.1% Sodium Azide. | |
| Rabbit | |
| Protein A purified | |
| Primary | |
| Human | |
| Purified |
| Immunocytochemistry | |
| Alexa Fluor 647 | |
| H2AFX; H2AX | |
| KLH-conjugated linear peptide corresponding to phospho H2A.X (Ser139). | |
| 100 μL | |
| NP_002096 | |
| 2-8°C for 12 months | |
| IgG |
For Research Use Only