missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Gab1 (Tyr627) Rabbit anti-Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 07833
Description
This Anti-phospho-Gab1 (Tyr627) Antibody is validated for use in western blotting for the detection of phospho-Gab1 (Tyr627).Specifications
| phospho-Gab1 (Tyr627) | |
| Polyclonal | |
| 100μL of affinity purified rabbit polyclonal IgG in 50% storage buffer (PBS (without Mg2+ and Ca2+), pH 7.3 containing 1.0mg/mL BSA (IgG, protease free) and 0.05% sodium azide) and 50% glycerol. | |
| Rabbit | |
| Affinity Purified | |
| Primary | |
| Mouse | |
| IgG |
| Western Blot | |
| Unconjugated | |
| GAB1 | |
| Peptide corresponding to amino acid region encompassing the human and mouse phospho-Gab1 (Tyr627) | |
| 100 μL | |
| NM_002039.2; NM_207123.1 | |
| -20°C in undiluted aliquotes for up to 24 months, Avoid Freeze/Thaw Cycles |
For Research Use Only