missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-erbB2 (Thr686) Mouse anti-Canine, Human, Mouse, Clone: 7F8, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 04 293
Description
Anti-phospho-erbB2 (Thr686) Antibody, clone 7F8 is an antibody against phospho-erbB2 (Thr686) for use in ELISA and western blotting.Specifications
| phospho-erbB2 (Thr686) | |
| Monoclonal | |
| Unconjugated | |
| NM_001005862.1; NM_004448.2 | |
| Mouse | |
| Subsequent thiophilic adsorption and size exclusion chromatography | |
| Primary | |
| −20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles. | |
| IgG1 |
| ELISA, Western Blot | |
| 7F8 | |
| In 1 mL of PBS containing 0.09% sodium azide, PEG, sucrose and 50% glycerol. | |
| ERBB2, HER2, HER-2, NGL, herstatin, erbB-2, C-erbB-2, NEU, TKR1, p185erbB2, HER-2/neu | |
| KLH-conjugated synthetic peptide encompassing the surrounding amino acids of Thr 686 in human erbB2 | |
| 100 μg | |
| Canine, Human, Mouse | |
| Purified |
For Research Use Only