missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-eIF4E (Ser209) Rabbit anti-Human, Polyclonal, MilliporeSigma™
Description
Anti-phospho eIF4E (Ser209) Antibody is an antibody against phospho eIF4E (Ser209) for use in western blotting.
Specifications
Specifications
| Antigen | phospho eIF4E (Ser209) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Gene Symbols | EIF4E; eIF4E; CBP; eIF-4E; MGC111573; EIF-4E; EIF4E1; EIF4F; EIF4EL1 |
| Host Species | Rabbit |
| Immunogen | KLH conjugated synthetic peptide corresponding to amino acids surrounding Ser209 of human eIF4E. Sequence conserved in mouse and rat. |
| Purification Method | Affinity Purified |
| Quantity | 100 μL |
| Primary or Secondary | Primary |
| Show More |
For Research Use Only