Learn More
Phospho-CREB (Ser133) Mouse anti-Mouse, Rat, Clone: 634-2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier:  MilliporeSigma 05807
Catalog No. 05807MI
Only null left
Description
Description
Use Anti-phospho-CREB (Ser133) Antibody, clone 634-2 (mouse monoclonal antibody) validated in immunocytochemistry, immunofluoresence, western blotting to detect phospho-CREB (Ser133) also known as active transcription factor CREB, cAMP responsive element binding protein 1.
Specifications
Specifications
phospho-CREB (Ser133)
Monoclonal
Unconjugated
NM_134442
Mouse
Protein G purfied
Primary
−20°C for up to 12 months.
IgG1 κ
Immunocytochemistry, Immunofluorescence, Western Blot
634-2
Purified mouse IgG1κ in buffer containing 0.014Mphosphate buffer, pH 7.6, 0.175MNaCl, 0.07% sodium azide and 30% glycerol. Liquid at −20deg;C.
CREB, MGC9284
A proprietary immunogen based on the amino acid sequence containing phospho-serine corresponding to residue 133 of human CREB.
200 μg
Mouse, Rat
Purified
Product Suggestions
Product Suggestions