missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-ATF2 (Thr69/71) Mouse anti-Human, Mouse, Clone: AW65, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05891
Description
Anti-phospho-ATF2 (Thr69/71) Antibody, clone AW65 is a high quality Mouse Monoclonal Antibody for the detection of phospho-ATF2 (Thr69/71) and has been validated in immunoprecipitation and western blotting.Specifications
| phospho-ATF2 (Thr69/71) | |
| Monoclonal | |
| Unconjugated | |
| ATF2, MGC111558, CREB2, TREB7, CRE-BP1, CREBP1, HB16 | |
| KLH-conjugated, synthetic peptide containing the sequence ...DQ[pT]P[pT]P... in which [pT] corresponds to phospho-threonine at residues 69 and 71 of human ATF2 | |
| 200 μg | |
| Human, Mouse | |
| Purified |
| Immunoprecipitation, Western Blot | |
| AW65 | |
| NM_001880.2 | |
| Mouse | |
| Protein G purfied | |
| Primary | |
| −20°C for up to 24 months. | |
| IgG1 |
For Research Use Only