missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-erbB2/Her2 (Tyr1112) Mouse anti-Canine, Human, Mouse, Rat, Clone: 19G5, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 04294
Description
Detect phosho-erbB2/Her2 (Tyr1112) using this Anti-phosho-erbB2/Her2 (Tyr1112) Antibody, clone 19G5 validated for use in ELISA and western blotting.Specifications
| phosho-erbB2/Her2 (Tyr1112) | |
| Monoclonal | |
| Unconjugated | |
| NM_001005862.1; NM_004448.2 | |
| Mouse | |
| Subsequent thiophilic adsorption and size exclusion chromatography | |
| Primary | |
| −20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles. | |
| IgG1 |
| ELISA, Western Blot | |
| 19G5 | |
| Lyophilized in 2X PBS containing 0.09% sodium azide, PEG, and sucrose. | |
| ERBB2, HER2, HER-2, NGL, herstatin, erbB-2, C-erbB-2, NEU, TKR1, p185erbB2, HER-2/neu | |
| KLH-conjugated synthetic peptide encompassing the surrounding amino acids of Tyr1112 in human erbB2/Her2 | |
| 100 μg | |
| Canine, Human, Mouse, Rat | |
| Purified |
For Research Use Only