missing translation for 'onlineSavingsMsg'
Learn More

prefoldin subunit 5, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004235
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89004235 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89004235 Supplier Abnova Corporation Supplier No. H00005204A01
Only null left

Mouse polyclonal antibody raised against a partial recombinant PFDN5.

This gene encodes a member of the prefoldin alpha subunit family. The encoded protein is one of six subunits of prefoldin, a molecular chaperone complex that binds and stabilizes newly synthesized polypeptides, thereby allowing them to fold correctly. The complex, consisting of two alpha and four beta subunits, forms a double beta barrel assembly with six protruding coiled-coils. The encoded protein may also repress the transcriptional activity of the proto-oncogene c-Myc. Alternatively spliced transcript variants encoding different isoforms have been described. [provided by RefSeq

Sequence: QTKYVEAKDCLNVLNKSNEGKELLVPLTSSMYVPGKLHDVEHVLIDVGTGYYVEKTAEDAKDFFKRKIDFLTKQMEKIQPALQEKHAMKQAVMEMMSQKI

Specifications

Antigen prefoldin subunit 5
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant PFDN5.
Formulation 50% glycerol
Gene PFDN5
Gene Accession No. NM_002624
Gene Alias MGC5329/MGC71907/MM-1/MM1/PFD5
Gene Symbols PFDN5
Host Species Mouse
Immunogen PFDN5 (NP_002615, 40 a.a. ∼ 139 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Transcription Regulation
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 5204
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less