missing translation for 'onlineSavingsMsg'
Learn More

PERP Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABE606MI
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE606MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE606MI Supplier MilliporeSigma Supplier No. ABE606
Only null left

Rabbit Polyclonal Antibody

Detect P53 apoptosis effector related to PMP-22 using this rabbit polyclonal antibody, Anti-PERP Antibody validated for use in western blotting and immunocytochemistry.

Specifications

Antigen PERP
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.
Gene Symbols PERP
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human PERP.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only