missing translation for 'onlineSavingsMsg'
Learn More
Learn More
poly(rC) binding protein 3, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Description
This gene encodes a member of the KH-domain protein subfamily. Proteins of this subfamily, also referred to as alpha-CPs, bind to RNA with a specificity for C-rich pyrimidine regions. Alpha-CPs play important roles in post-transcriptional activities and have different cellular distributions. This gene's protein is found in the cytoplasm, yet it lacks the nuclear localization signals found in other subfamily members. Alternative splicing results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq]
Sequence: MESKVSEGGLNVTLTIRLLMHGKEVGSIIGKKGETVKKMREESGARINISEGNCPERIVTITGPTDAIFKAFAMIAYKFEEDIINSMSNSPATSKPPVTLRLVVPASQCGSLIGKGGSKIKEIRESTGAQVQVAGDMLPNSTERAVTISGTPDAIIQCVKQICVVMLEAYTIQGQYAIPHPDLTKLHQLAMQQTPFPPLGQTNPAFPGEKLPLHSSEEAQNLMGQSSGLDASPPASTHELTIPNDLIGCIIGRQGTKINEIRQMSGAQIKIANATEGSSERQITITGTPANISLAQYLINARLTSEVTGMGTL
Specifications
Specifications
| Antigen | poly(rC) binding protein 3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Mouse polyclonal antibody raised against a full-length human PCBP3 protein. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | PCBP3 |
| Gene Accession No. | BC012061.1 |
| Gene Alias | ALPHA-CP3 |
| Gene Symbols | PCBP3 |
| Show More |