missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARP Poly ADP-ribose Polymerase-1 Rabbit anti-Bovine, Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB16661
Description
This Anti-PARP Poly ADP-ribose Polymerase-1 Antibody is validated for use in ELISA, western blotting for the detection of PARP Poly ADP-ribose Polymerase-1.Specifications
| PARP Poly ADP-ribose Polymerase-1 | |
| Polyclonal | |
| Antiserum containing 0.05% sodium azide. Liquid containing 0.05% sodium azide. | |
| Rabbit | |
| 100 μL | |
| NM_001618.3 | |
| −20°C in undiluted aliquotes for 12 months. |
| ELISA, Western Blot | |
| Unconjugated | |
| PARP1; poly(ADP-ribosyl)transferase; pADPRT-1; PARP-1; ADPRT1; Poly; PARP; ADPRT; PPOL; NAD; EC 2.4.2.30 | |
| Synthetic peptide based on the sequence from the C-terminal portion of the bovine PARP automodification domain (amino acids 509-524) conjugated to a carrier. | |
| Primary | |
| Bovine, Human, Mouse, Rat | |
| Serum |
For Research Use Only