missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pannexin 1 Rabbit anti-Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Invitrogen 488000
Description
The pannexin gene family encodes a second class of putative gap junction proteins and are highly conserved in invertebrates and mammals. Pannexins (Panx) are four-pass transmembrane proteins that oligomerize to form large pore ion and metabolite-permeable channels. Pannexin-1 (PANX1) and Pannexin-3 are closely related, while Pannexin-2 is a more distant relation. PANX1 is a transmembrane protein that forms a mechanosensitive ATP-permeable channel between adjacent cells and in the endoplasmic reticulum. PANX1 may play a role as a Ca2+ -leak channel to regulate ER Ca2+ homeostasis and regulates neural stem and progenitor cell proliferation.Specifications
| Pannexin 1 | |
| Polyclonal | |
| Unconjugated | |
| Q9JIP4, P60570 | |
| Panx1 | |
| Synthetic peptide derived from the internal region of the mouse Pannexin 1 protein (AccessionNo. NP_062355), which is identical to rat and human sequence. | |
| 100 μg | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry, Western Blot | |
| 0.25 mg/mL | |
| Panx1 | |
| PANX1, Pannexin 1, Pannexin-1, PX1, MRS1, UNQ2529, UNQ2529/PRO6028 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 315435, 55991 | |
| -20°C | |
| Liquid |