Learn More
NR1 Mouse anti-Human, Rat, Mouse, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier:  MilliporeSigma 05432
Catalog No. 05432MI
Only null left
Explore available promotions
Description
Description
Anti-NR1 Antibody, CT detects level of NR1 and has been published and validated for use in immunocytochemistry, immunoprecipitation and western blotting.
Specifications
Specifications
NR1
Monoclonal
Purified mouse monoclonal IgG in buffer containing 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide.
NR1, Grin1, Nmdar1
His-tag fusion protein corresponding to residues 834-938 of rat NR1A. The immunizing sequence is identical in rat and human.
200 μg
Human, Mouse, Rat
Purified
Immunocytochemistry, Immunoprecipitation, Western Blot
Unconjugated
NM_017010
Mouse
Protein G purfied
Primary
2-8°C for up to 12 months.
IgG
Product Suggestions
Product Suggestions