missing translation for 'onlineSavingsMsg'
Learn More

Norepinephrine Transporter Mouse anti-Human, Monkey, MilliporeSigma™

Catalog No. MAB5620MI
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5620MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5620MI Supplier MilliporeSigma Supplier No. MAB5620

Mouse Monoclonal Antibody

Detect Norepinephrine Transporter using this Anti-Norepinephrine Transporter Antibody validated for use in immunohistochemistry.

Specifications

Antigen Norepinephrine Transporter
Applications Immunohistochemistry
Classification Monoclonal
Conjugate Unconjugated
Formulation Mouse ascites. Liquid. Contains no preservative.
Gene Accession No. NM_001043.2
Gene Symbols SLC6A2, NET, SLC6A5, NAT1, NET1
Host Species Mouse
Immunogen Synthetic peptide from human norepinephrine Transporter.
Purification Method Ascites
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Monkey
Content And Storage −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG
Show More Show Less

For Research Use Only