missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NMDAR2B phospho Tyrosine 1336 Rabbit anti-Rat, Polyclonal, MilliporeSigma™
Description
Detect NMDAR2B phospho Tyrosine 1336 using this Anti-NMDAR2B phospho Tyrosine 1336 Antibody validated for use in western blotting.
Specifications
Specifications
| Antigen | NMDAR2B phospho Tyrosine 1336 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Liquid in 10mM HEPES (pH 7.5), 150mM NaCl, 100μg/mL BSA and 50% glycerol. |
| Gene Symbols | GRIN2B; MGC142180; Glutamate; NMDAR2B; NR3; NR2B; MGC142178; hNR3 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide of amino acids surrounding the phosphoTyrosine 1336 site of rat NMDAR2B. The immunogen has 100% similarity with human and mouse. |
| Purification Method | Affinity Purified |
| Quantity | 100 μL |
| Show More |
For Research Use Only