missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ninein Mouse anti-Human, Clone: 79-160-7, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABT29
Description
Anti-Ninein Antibody, clone 79-160-7 is an antibody against Ninein for use in western blotting and immunoprecipitation.Specifications
| Ninein | |
| Monoclonal | |
| Unconjugated | |
| NP_891989 | |
| Mouse | |
| Protein G purfied | |
| Primary | |
| 2-8°C for one year | |
| IgG2a κ |
| Immunoprecipitation, Western Blot | |
| 79-160-7 | |
| Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine, pH 7.4, 150mM NaCl with 0.05% sodium azide. | |
| ninein, hNinein, NIN | |
| Recombinant protein corresponding to human Ninein. | |
| 100 μg | |
| Human | |
| Purified |
For Research Use Only