missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NeuN Mouse anti-Human, Phycoerythrin, Clone: A60, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma FCMAB317PE
Description
This Milli-Mark™ Anti-NeuN-PE Antibody, clone A60 is validated for use in FC for the detection of NeuN.Specifications
| NeuN | |
| Monoclonal | |
| PE | |
| NeuN, A58 | |
| Purified cell nuclei from mouse brain. | |
| 100 Tests | |
| Human | |
| Purified |
| Flow Cytometry | |
| A60 | |
| Purified mouse monoclonal IgG1 conjugated to PE in PBS with 0.1% sodium azide and 15mg/mL BSA | |
| Mouse | |
| Protein A Purified | |
| Primary | |
| 2-8°C in undiluted aliquots for up to 6 months. Protect from Light. | |
| IgG1 |
For Research Use Only