missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nerve Growth Factor Receptor Mouse anti-Human, Pig, Rabbit, Sheep, AlexaFluor™488, Clone: ME20.4, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB5386X
Description
This Anti-Nerve Growth Factor Receptor Antibody, p75, Alexa Fluor 488 is validated for use in flow cytometry, IC for the detection of Nerve Growth Factor Receptor.Specifications
| Nerve Growth Factor Receptor | |
| Monoclonal | |
| Alexa Fluor 488 | |
| NM_002507.2 | |
| Mouse | |
| 50 μg | |
| Human, Pig, Rabbit, Sheep | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry | |
| ME20.4 | |
| Liquid. Contains no preservative. | |
| NGFR, p75(NTR), TNFRSF16, Gp80-LNGFR, CD271 | |
| Human melanoma cell line. | |
| Primary | |
| −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. |
For Research Use Only