missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NckΒ Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Description
This Anti-Nckβ Antibody is validated for use in immunoprecipitation, western blotting for the detection of Nckβ.
Specifications
Specifications
| Antigen | Nckβ |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30% |
| Gene Symbols | NCK2; NCKbeta; NCKBETA; GRB4; Nck-2 |
| Host Species | Rabbit |
| Immunogen | peptide corresponding to amino acids 175-190 of murine Nckβ (EAPSFLSLRRGTALSN) |
| Purification Method | Protein A chromatography |
| Quantity | 200 μg |
| Show More |
For Research Use Only