missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-CoR Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABE251
Description
Anti-N-CoR Antibody is a Rabbit Polyclonal Antibody for detection of N-CoR also known as Nuclear receptor corepressor 1 and has been validated in western blotting.Specifications
| N-CoR | |
| Polyclonal | |
| Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
| Rabbit | |
| Affinity Purified | |
| Primary | |
| Human, Mouse |
| Western Blot | |
| Unconjugated | |
| Ncor1; Rxrip13 | |
| KLH-conjugated linear peptide corresponding to a region between amino acids 1510 and 1540 of the N-CoR protein. | |
| 100 μg | |
| NP_035438 | |
| 2-8°C for 12 months |
For Research Use Only