missing translation for 'onlineSavingsMsg'
Learn More

Myosin XIX (Myo19) Rabbit anti-Human, Canine, Polyclonal, MilliporeSigma™

Catalog No. ABT41MI
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABT41MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABT41MI Supplier MilliporeSigma Supplier No. ABT41
Only null left

Rabbit Polyclonal Antibody

Anti-Myosin XIX (Myo19) Antibody detects level of Myosin XIX (Myo19) and has been published and validated for use in immunohistochemistry (paraffin).

Specifications

Antigen Myosin XIX (Myo19)
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols MYO19; MYOHD1
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human Myosin XIX (Myo19).
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_001028752
Target Species Canine, Human
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only