missing translation for 'onlineSavingsMsg'
Learn More

Muscarinic Acetylcholine Receptor m2 Rat anti-Human, Mammals, Monkey, Rat, Clone: M2-2-B3, MilliporeSigma™

Catalog No. MAB367MI
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB367MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB367MI Supplier MilliporeSigma Supplier No. MAB367
Only null left

Rat Monoclonal Antibody

Anti-Muscarinic Acetylcholine Receptor m2 Antibody, clone M2-2-B3 is an antibody against Muscarinic Acetylcholine Receptor m2 for use in immunocytochemistry, immunohistochemistry and immunoprecipitation.

Specifications

Antigen Muscarinic Acetylcholine Receptor m2
Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone M2-2-B3
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in 0.02Mphosphate buffer, 0.25MNaCl with 0.1% sodium azide, pH 7.6.
Gene Accession No. NM_000739.2; NM_001006626.1; NM_001006627.1; NM_001006628.1; NM_001006629.1; NM_001006630.1; NM_001006631.1; NM_001006632.1
Gene Symbols CHRM2, MGC120006, FLJ43243, MGC120007, HM2
Host Species Rat
Immunogen i3 loop of m2 receptor fusion protein (225-359), fused to Glutathione S-transferase.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Monkey, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only