missing translation for 'onlineSavingsMsg'
Learn More

anti-Mouse IgG peroxidase-linked whole antibody (from sheep) Secondary Antibody, Cytiva

Catalog No. 45000692
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45000692 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45000692 Supplier Cytiva Supplier No. NXA9311ML
Only null left

Highly species specific HRP-conjugated antibodies optimized for use with Amersham ECL Western Blotting Detection Reagents.

Host Species: Sheep
Isotype: IgG
Species Reactivity: Ms

Protein Blotting, ELISA, Immunocytochemistry

Specifications

Antigen IgG, Whole Molecule
Classification Monoclonal
Conjugate HRP
Host Species Sheep
Purification Method Affinity Purified
Quantity 1 mL
Primary or Secondary Secondary
Isotype IgG