missing translation for 'onlineSavingsMsg'
Learn More

anti-Mouse IgG, peroxidase-linked species-specific whole antibody (from sheep) Secondary Antibody, Cytiva

Catalog No. 45001275
Change view
Click to view available options
Quantity:
0.1 mL
1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45001275 0.1 mL
45000679 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45001275 Supplier Cytiva Supplier No. NA931100UL
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Highly species-specific anti-Mouse IgG. Cytiva Lifescience™ anti-Mouse IgG, peroxidase-linked species-specific whole antibody (from sheep) Secondary Antibody is designed for applications such as protein blotting, ELISA and immunocytochemistry.

Host Species: Sheep
Isotype: IgG
Species Reactivity: Ms

 

Specifications

Antigen IgG, Whole Molecule
Classification Monoclonal
Conjugate HRP
Host Species Sheep
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Secondary
Isotype IgG