missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Mouse IgG, peroxidase-linked species-specific F(ab')2 fragment (from sheep) Secondary Antibody, Cytiva

Use for protein blotting, ELISA and immunocytochemistry applications.  Anti-Mouse IgG, peroxidase-linked species-specific F(ab')2 fragment (from sheep) Secondary Antibody, Cytiva, is a secondary antibody hosted in sheep.  

Supplier:  Cytiva NA93101ML

Catalog No. 45-000-680



Description

Description

Host Species: Sheep
Isotype: IgG
Species Reactivity: Ms

Protein Blotting, ELISA, Immunocytochemistry

Specifications

Specifications

IgG F(Ab')2
HRP
Affinity Purified
Secondary
Monoclonal
Sheep
1 mL
IgG
SDS
Documents

Documents

Promotions

Promotions