missing translation for 'onlineSavingsMsg'
Learn More

Monomethyl-Histone H3 (Lys27) Rabbit anti-Human, Vertebrates, Polyclonal, MilliporeSigma™

Catalog No. 07448SMI
Change view
Click to view available options
Quantity:
20 μg
200 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07448SMI 20 μg
07448MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 07448SMI Supplier MilliporeSigma Supplier No. 07448S
Only null left

Rabbit Polyclonal Antibody

Anti-monomethyl-Histone H3 (Lys27) Antibody is a rabbit polyclonal antibody to detect Histone H3 monomethylated at lysine 27. This purified Ab, also known as Anti-H3K27me1 is specificity verified by dot blot (DB), published in peer reviewed journals and validated in immunocytochemistry, western blot, Chimmunoprecipitation, ChIP-seq.

Specifications

Antigen monomethyl-Histone H3 (Lys27)
Applications ChIP sequencing (ChIP-seq), Dot Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit IgG in buffer containing 0.01M phosphate buffer, pH 7.6, 0.125M NaCl, 0.05% sodium azide and 30% glycerol.
Gene Symbols HIST1H3A; H3FA; HIST1H3B; H3FL; HIST1H3C; H3FC; HIST1H3D; H3FB; HIST1H3E; H3FD; HIST1H3F; H3FI; HIST1H3G; H3FH; HIST1H3H; H3FK; HIST1H3I; H3FF; HIST1H3J; H3FJ; HIST3H3; H3FT; H3T; MGC126886
Host Species Rabbit
Immunogen KLH-conjugated, synthetic 2X-branched peptide containing the sequence ...ARmeKSA... in which meK corresponds to monomethyl-lysine at residue 27 of human histone H3.
Purification Method Protein A purfied
Quantity 20 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_003484
Target Species Human, Vertebrates
Content And Storage 2-8°C for 12 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only