missing translation for 'onlineSavingsMsg'
Learn More

MMP-1 Mouse anti-Human, Rabbit, Clone: 41-1E5, MilliporeSigma™

Catalog No. mab3307mi
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3307MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3307MI Supplier MilliporeSigma Supplier No. MAB3307
Only null left

Mouse Monoclonal Antibody

Anti-MMP-1 Antibody, a.a. 332-350 hMMP1, clone 41-1E5 detects level of MMP-1 and has been published and validated for use in ELISA, immunohistochemistry (paraffin) and western blotting.

Specifications

Antigen MMP-1
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 41-1E5
Conjugate Unconjugated
Formulation Liquid in 0.1MNa-Phosphate buffer, pH 7.0 containing 2% protease free bovine Serum albumin.
Gene Accession No. NM_002421.2
Gene Symbols MMP1, CLG, MMP-1, CLGN, EC 3.4.24.7 [Contains: 22 kDa interstitial collagenase, 27 kDa interstitial collagenase].
Host Species Mouse
Immunogen Syntheic peptide aa332-350 of hMMP-1
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Rabbit
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only