missing translation for 'onlineSavingsMsg'
Learn More

Metabotropic Glutamate Receptor 6 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ab9450mi
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB9450MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB9450MI Supplier MilliporeSigma Supplier No. AB9450
Only null left

Rabbit Polyclonal Antibody

This Anti-Metabotropic Glutamate Receptor 6 Antibody is validated for use in immunohistochemistry (paraffin) for the detection of Metabotropic Glutamate Receptor 6.

Specifications

Antigen Metabotropic Glutamate Receptor 6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified immunoglobulin. Liquid in PBS, pH 7.7 with 0.01% sodium azide.
Gene Symbols GRM6; DKFZp686H1993; GPRC1F; mGluR6; MGLUR6; CSNB1B; mGlu6
Host Species Rabbit
Immunogen Synthetic peptide from the N-terminal extracellular domain of human Metabotropic Glutamate Receptor 6.
Purification Method Affinity Purified
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_000843.3
Target Species Human
Content And Storage −70°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only