missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAML1 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB5975
Description
Detect MAML1 using this Anti-MAML1 Antibody validated for use in western blot, immunohistochemistry.Specifications
| MAML1 | |
| Polyclonal | |
| Affinity purified immunoglobulin. Precipitated antibody in a solution of 50% saturated ammonium sulfate and PBS containing no preservatives. | |
| Rabbit | |
| Affinity Purified | |
| Primary | |
| Human, Mouse |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| MAML1; MAM1; KIAA0200; Mam1; Mam-1 | |
| Synthetic peptide, amino acids 661-676 of MAML1. | |
| 100 μg | |
| NM_014757.3 | |
| -70°C for up to 6 months, Avoid Freeze/Thaw Cycles |
For Research Use Only