Learn More
MAGI2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant MAGI2.
Supplier: Abnova Corporation H00009863A01
Description
The protein encoded by this gene interacts with atrophin-1. Atrophin-1 contains a polyglutamine repeat, expansion of which is responsible for dentatorubral and pallidoluysian atrophy. This encoded protein is characterized by two WW domains, a guanylate kinase-like domain, and multiple PDZ domains. It has structural similarity to the membrane-associated guanylate kinase homologue (MAGUK) family. [provided by RefSeq
Sequence: PANSMVPPLAIMERPPPVMVNGRHNYETYLEYISRTSQSVPDITDRPPHSLHSMPTDGQLDGTYPPPVHDDNVSMASSGATQAELMTLTIVKGAQGFGFTIADSPTGQRVSpecifications
| MAGI2 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant MAGI2. | |
| MAGI2 | |
| ACVRIP1/AIP1/ARIP1/MAGI-2/SSCAM | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_012301 | |
| MAGI2 | |
| MAGI2 (NP_036433, 519 a.a. ∼ 628 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 9863 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |