missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lipocalin-2 Mouse anti-Human, Clone: PA348-26.3.5, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABN481
Description
Detect Lipocalin using this mouse monoclonal antibody, Anti-Lipocalin-2 Antibody, clone PA348-26.3.5 validated for use in western blotting, immunohistochemistry and ELISA.Specifications
| Lipocalin-2 | |
| Monoclonal | |
| Unconjugated | |
| NP_005555.1 | |
| Mouse | |
| Protein G purified | |
| Primary | |
| 2-8°C for one year | |
| IgG1 κ |
| ELISA, Immunohistochemistry, Western Blot | |
| PA348-26.3.5 | |
| Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
| LCN2, HNL, NGAL | |
| Recombinant protein corresponding to human Lipocalin-2. | |
| 100 μg | |
| Human | |
| Purified |
For Research Use Only