missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Lewis Y Monoclonal Antibody , TCI America™
Anti-Lewis Y Monoclonal Antibody, TCI America™
Supplier: TCI America A25101VIAL
Specifications
| Lewis Y | |
| Monoclonal | |
| Mouse | |
| Primary | |
| Purified |
| ELISA, Flow Cytometry, Immunohistochemistry, Western Blot, TLC-Immunostaining | |
| H18A | |
| Immunized with human gastric cancer cells (MKN74), and screened using cancer glycolipids | |
| Store at -20°C to -80°C (preferably at -80°C). It is not recommended storing at 4°C. Avoid repeated freezing and thawing. | |
| IgG3 κ |